Anti-ERO1A Rabbit Polyclonal Antibody
Catalog # 77736-145
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ERO1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:Q96HE7
- Antigen Synonyms:Endoplasmic oxidoreductin 1 like protein|Endoplasmic oxidoreductin-1-like protein|Endoplasmic reticulum oxidoreductase 1 alpha|Endoplasmic reticulum oxidoreductin 1-like|ERO1 alpha|ERO1 L|ERO1 Lalpha|ERO1 like protein alpha|ERO1-alpha|ERO1-L|ERO1-L-alpha|ERO1-like (S. cerevisiae)|ERO1-like alpha|ERO1-like protein alpha|ERO1-like|S. cerevisiae|homolog of|alpha|ERO1A_HUMAN|ERO1L|ERO1LA|Oxidoreductin 1 Lalpha|Oxidoreductin-1-L-alpha|PRO865|UNQ434
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:ERO1L
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ERO1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse