Anti-EXOC6 Rabbit Polyclonal Antibody
77736-282
New Product
- Antigen Symbol:EXOC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8TAG9
- Antigen Synonyms:EXOC6|SEC15A|SEC15L|SEC15L1|Exocyst complex component 6|Exocyst complex component Sec15A|SEC15-like protein 1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:SEC15-like protein 1
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: EXOC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat