Anti-INPP4B Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INPP4B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77741-096
- 77858-122
- Antigen Symbol:
- INPP4B
- INPP4B
- Antigen Synonyms:
- Inositol polyphosphate 4 phosphatase|type II|4-bisphosphate 4-phosphatase|Inositol polyphosphate 4 phosphatase type II 105kDa|Inositol polyphosphate 4-phosphatase type II|INP4B_HUMAN|MGC132014|Type II inositol 3,4 bisphosphate 4 phosphatase|Type II inositol-3
- Inositol polyphosphate 4 phosphatase|type II|4-bisphosphate 4-phosphatase|Inositol polyphosphate 4 phosphatase type II 105kDa|Inositol polyphosphate 4-phosphatase type II|INP4B_HUMAN|MGC132014|Type II inositol 3,4 bisphosphate 4 phosphatase|Type II inositol-3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15327
- O15327
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Type II inositol 3,4-bisphosphate 4-phosphatase protein (1-53AA)
- Recombinant Human Type II inositol 3,4-bisphosphate 4-phosphatase protein (1-53AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice