Anti-GRB10 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: GRB10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77856-417
- 77738-977
- Antigen Symbol:
- GRB10
- Antigen Synonyms:
- grb 10|GRB IR|grb-10|GRB10 adapter protein|GRB10 adaptor protein|GRB10|GRB10_HUMAN|GRBIR|Growth factor receptor bound protein 10|Growth factor receptor-bound protein 10|Insulin receptor binding protein|Insulin receptor binding protein GRB IR|Insulin receptor-binding protein Grb-IR|IRBP|KIAA0207|Maternally expressed gene 1|MEG1|RSS
- grb 10|GRB IR|grb-10|GRB10 adapter protein|GRB10 adaptor protein|GRB10|GRB10_HUMAN|GRBIR|Growth factor receptor bound protein 10|Growth factor receptor-bound protein 10|Insulin receptor binding protein|Insulin receptor binding protein GRB IR|Insulin receptor-binding protein Grb-IR|IRBP|KIAA0207|Maternally expressed gene 1|MEG1|RSS
- Clonality:
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- Q13322
- Q13322
- Isotype:
- IgG
- ELISA:
- Yes
- ImmunoChemistry:
- Yes
- ImmunoFluorescence:
- Yes
- Immunogen:
- Recombinant Human Growth factor receptor-bound protein 10 protein (15-180AA)
- Recombinant Human Growth factor receptor-bound protein 10 protein (15-180AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice