Anti-FOXA1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77737-396
- 77855-533
- Antigen Symbol:
- FOXA1
- FOXA1
- Antigen Synonyms:
- forkhead box A1|Forkhead box protein A1|FOX A1|FOXA1_HUMAN|hepatocyte nuclear factor 3 alpha|Hepatocyte nuclear factor 3-alpha|HNF 3A|HNF-3-alpha|HNF-3A|HNF3A|MGC33105|TCF 3A|TCF-3A|TCF3A|Transcription factor 3A
- forkhead box A1|Forkhead box protein A1|FOX A1|FOXA1_HUMAN|hepatocyte nuclear factor 3 alpha|Hepatocyte nuclear factor 3-alpha|HNF 3A|HNF-3-alpha|HNF-3A|HNF3A|MGC33105|TCF 3A|TCF-3A|TCF3A|Transcription factor 3A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55317
- P55317
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hepatocyte nuclear factor 3-alpha protein (367-472AA)
- Recombinant Human Hepatocyte nuclear factor 3-alpha protein (367-472AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice