Anti-CXCR2 Rabbit Recombinant polyclonal Antibody
New Product
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: CXCR2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77858-087
- 77741-043
- Antigen Symbol:
- CXCR2
- CXCR2
- Antigen Synonyms:
- C-X-C chemokine receptor type 2|CD 182|CD182|CD182 antigen|CDw128b|Chemokine (CXC) receptor 2|CMKAR2|CXC-R2|CXCR 2|CXCR-2|CXCR2_HUMAN|GRO/MGSA receptor|High affinity interleukin-8 receptor B|IL 8 receptor type 2|IL 8R B|IL-8 receptor type 2|IL-8R B|IL8 RB|IL8 receptor type 2|IL8R B|IL8R2|IL8RA|Interleukin 8 Receptor B|Interleukin 8 receptor|beta|type 2
- C-X-C chemokine receptor type 2| CD 182| CD182| CD182 antigen| CDw128b| Chemokine (CXC) receptor 2| CMKAR2| CXC-R2| CXCR 2| CXCR-2| CXCR2_HUMAN| GRO/MGSA receptor| High affinity interleukin-8 receptor B| IL 8 receptor type 2| IL 8R B| IL-8 receptor type 2| IL-8R B| IL8 RB| IL8 receptor type 2| IL8R B| IL8R2| IL8RA| Interleukin 8 Receptor B| Interleukin 8 receptor| beta| type 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25025
- P25025
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human C-X-C chemokine receptor type 2 protein (1-40AA)
- Recombinant Human C-X-C chemokine receptor type 2 protein (1-40AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice