Anti-MCM7 Rabbit Polyclonal Antibody
Catalog # 77742-916
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MCM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:P33993
- Antigen Synonyms:CDABP0042|CDC 47|CDC47|CDC47 homolog|Cdc47|S. cerevisiae|homolog of|DNA replication licensing factor MCM7|Homolog of S. cerevisiae Cdc47|MCM 2|MCM 7|MCM2|formerly|Mcm7|MCM7 minichromosome maintenance deficient 7|MCM7_HUMAN|Minichromosome Maintainence 7|Minichromosome maintainence|Minichromosome maintenance complex component 7|Minichromosome maintenance deficient 7|Minichromosome maintenance protein 7|P1.1 MCM3|P1.1-MCM3|P1CDC47|P85MCM|PNAS 146|PNAS146|PPP1R104|Protein phosphatase 1|regulatory subunit 104
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:regulatory subunit 104
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MCM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat