Anti-FABP7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FABP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77736-422
- 77855-100
- Antigen Symbol:
- FABP7
- FABP7
- Antigen Synonyms:
- B FABP|B-FABP|BFABP|BLBP|Brain lipid binding protein|Brain lipid-binding protein|Brain-type fatty acid-binding protein|DKFZp547J2313|FABP 7|FABP7|FABP7_HUMAN|FABPB|Fatty Acid Binding Protein 7|Fatty acid binding protein 7 brain|Fatty acid binding protein brain|Fatty acid-binding protein 7|Fatty acid-binding protein, brain|Mammary derived growth inhibitor related|Mammary-derived growth inhibitor related|MRG|OTTHUMP00000017119
- B FABP| B-FABP| BFABP| BLBP| Brain lipid binding protein| Brain lipid-binding protein| Brain-type fatty acid-binding protein| DKFZp547J2313| FABP 7| FABP7| FABP7_HUMAN| FABPB| Fatty Acid Binding Protein 7| Fatty acid binding protein 7 brain| Fatty acid binding protein brain| Fatty acid-binding protein 7| Fatty acid-binding protein, brain| Mammary derived growth inhibitor related| Mammary-derived growth inhibitor related| MRG| OTTHUMP00000017119
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15540
- O15540
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fatty acid-binding protein, brain protein (2-132AA)
- Recombinant Human Fatty acid-binding protein, brain protein (2-132AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice