Anti-NRBP2 Rabbit Polyclonal Antibody
77745-007
New Product
- Antigen Symbol:NRBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NSY0
- Antigen Synonyms:BC011468|DKFZp434P086|MGC12117|MGC138699|MGC18994|NRBP2_HUMAN|Nuclear receptor binding protein 2|Nuclear receptor-binding protein 2|PP9320|Transformation related gene 16 protein|Transformation-related gene 16 protein|transformation-related protein 16|TRG 16|TRG-16|TRG16
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:TRG16
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NRBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat