Anti-KCNN4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: KCNN4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-463
- 77741-524
- Antigen Symbol:
- KCNN4
- KCNN4
- Antigen Synonyms:
- DHS2|hIKCa1|hKCa4|hSK4|IK|IK1|IKCa1|Intermediate conductance calcium activated potassium channel protein 4|Intermediate conductance calcium-activated potassium channel protein 4|KCa3.1|KCa4|KCNN 4|KCNN4_HUMAN|Potassium calcium activated channel subfamily N member 4|Potassium intermediate/small conductance calcium activated channel, subfamily N, member 4|Putative Gardos channel|SK4|SKCa 4|SKCa4
- DHS2|hIKCa1|hKCa4|hSK4|IK|IK1|IKCa1|Intermediate conductance calcium activated potassium channel protein 4|Intermediate conductance calcium-activated potassium channel protein 4|KCa3.1|KCa4|KCNN 4|KCNN4_HUMAN|Potassium calcium activated channel subfamily N member 4|Potassium intermediate/small conductance calcium activated channel, subfamily N, member 4|Putative Gardos channel|SK4|SKCa 4|SKCa4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15554
- O15554
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Intermediate conductance calcium-activated potassium channel protein 4 protein (288-427AA)
- Recombinant Human Intermediate conductance calcium-activated potassium channel protein 4 protein (288-427AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice