Anti-NANS Rabbit Polyclonal Antibody
77743-879
New Product
- Antigen Symbol:NANS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9NR45
- Antigen Synonyms:EC 2.5.1.56|EC 2.5.1.57|N acetylneuraminate 9 phosphate synthase|N acetylneuraminate synthase|N acetylneuraminic acid phosphate synthase|N acetylneuraminic acid synthase|N-acetylneuraminate synthase|N-acetylneuraminate-9-phosphate synthase|N-acetylneuraminic acid phosphate synthase|N-acetylneuraminic acid synthase|OTTHUMP00000021769|SAS|Sialic acid phosphate synthase|Sialic acid synthase|SIAS_HUMAN
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:SIAS_HUMAN
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: NANS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat