Anti-F13B Rabbit Polyclonal Antibody
77736-611
New Product
- Antigen Symbol:F13B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P05160
- Antigen Synonyms:Coagulation factor XIII B chain|Fibrin-stabilizing factor B subunit|Protein-glutamine gamma-glutamyltransferase B chain|Transglutaminase B chain
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Transglutaminase B chain
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: F13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human