Anti-HOXD3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77857-611
- 77740-333
- Antigen Symbol:
- HOXD3
- HOXD3
- Antigen Synonyms:
- Homeo box D3|Homeobox D3|Homeobox protein Hox D3|Homeobox protein Hox-4A|Homeobox protein Hox-D3|Homeobox protein Homeodomain protein|HOX 1D|HOX 4|Hox 4.1|Hox 4A|HOX D3|Hox-4.1 mouse homolog of|HOX1D|HOX4|Hox4.1|HOX4A|HOXD 3|HXD3_HUMAN|MGC10470
- Homeo box D3|Homeobox D3|Homeobox protein Hox D3|Homeobox protein Hox-4A|Homeobox protein Hox-D3|Homeobox protein Homeodomain protein|HOX 1D|HOX 4|Hox 4.1|Hox 4A|HOX D3|Hox-4.1 mouse homolog of|HOX1D|HOX4|Hox4.1|HOX4A|HOXD 3|HXD3_HUMAN|MGC10470
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31249
- P31249
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Homeobox protein Hox-D3 protein (263-346AA)
- Recombinant human Homeobox protein Hox-D3 protein (263-346AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice