Anti-KIAA1191 Rabbit Polyclonal Antibody
77741-608
New Product
- Antigen Symbol:KIAA1191
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96A73
- Antigen Synonyms:Brain-derived rescue factor p60MONOX|Flavin monooxygenase motif-containing protein of 33 kDa|FLJ21022|Hypothetical protein LOC57179|P33MX_HUMAN|P60MONOX|Putative monooxygenase p33MONOX
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Putative monooxygenase p33MONOX
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: KIAA1191
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse