Anti-MRPL24 Rabbit Polyclonal Antibody
Catalog # 77743-405
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRPL24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q96A35
- Antigen Synonyms:39S ribosomal protein L24|mitochondrial|L24mt|MRP-L24|Mitochondrial large ribosomal subunit protein uL24m
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:mitochondrial large ribosomal subunit protein uL24m
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MRPL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat