Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:100 to 1:300
Type:
Antigen: DDX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77827-238
- 77827-239
- Antigen Symbol:
- DDX58
- DDX58
- Antigen Name:
- Synthetic peptide of DDX58
- Synthetic peptide of DDX58
- Antigen Synonyms:
- DDX58_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 58|DEAD (Asp Glu Ala Asp/His) box polypeptide|DEAD box protein 58|DEAD/H (Asp Glu Ala Asp/His) box polypeptide RIG1|DKFZp434J1111|DKFZp686N19181|FLJ13599|Probable ATP dependent RNA helicase Probable ATP-dependent RNA helicase Retinoic acid inducible gene 1 protein|Retinoic acid-inducible gene 1 protein|Retinoic acid-inducible gene I protein|RIG I|Rig-1|RIG-I|RIG1|rigi|RLR 1|RNA helicase|RNA helicase RIG I|SGMRT2
- DDX58_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 58|DEAD (Asp Glu Ala Asp/His) box polypeptide|DEAD box protein 58|DEAD/H (Asp Glu Ala Asp/His) box polypeptide RIG1|DKFZp434J1111|DKFZp686N19181|FLJ13599|Probable ATP dependent RNA helicase Probable ATP-dependent RNA helicase Retinoic acid inducible gene 1 protein|Retinoic acid-inducible gene 1 protein|Retinoic acid-inducible gene I protein|RIG I|Rig-1|RIG-I|RIG1|rigi|RLR 1|RNA helicase|RNA helicase RIG I|SGMRT2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95786
- O95786
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice