Anti-KANSL1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: KANSL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-065
- 77832-066
- Antigen Symbol:
- KANSL1
- KANSL1
- Antigen Name:
- KAT8 regulatory NSL complex subunit 1 protein (815-1086AA)
- KAT8 regulatory NSL complex subunit 1 protein (815-1086AA)
- Antigen Synonyms:
- CENP-36|centromere protein 36|DKFZp686P06109|DKFZp727C091|hMSL1v1|hypothetical protein LOC284058|K1267_HUMAN|KANSL1|KAT8 regulatory NSL complex subunit 1|KDVS|KIAA1267|male-specific lethal 1 homolog|MGC102843|MLL1/MLL complex subunit KIAA1267|MSL1 homolog 1|MSL1v1|Non specific lethal 1 homolog|NSL complex protein NSL1|NSL1
- CENP-36|centromere protein 36|DKFZp686P06109|DKFZp727C091|hMSL1v1|hypothetical protein LOC284058|K1267_HUMAN|KANSL1|KAT8 regulatory NSL complex subunit 1|KDVS|KIAA1267|male-specific lethal 1 homolog|MGC102843|MLL1/MLL complex subunit KIAA1267|MSL1 homolog 1|MSL1v1|Non specific lethal 1 homolog|NSL complex protein NSL1|NSL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z3B3
- Q7Z3B3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice