Anti-TRIM69 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TRIM69
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-746
- 77832-745
- Antigen Symbol:
- TRIM69
- TRIM69
- Antigen Name:
- E3 ubiquitin-protein ligase TRIM69 protein (1-341AA)
- E3 ubiquitin-protein ligase TRIM69 protein (1-341AA)
- Antigen Synonyms:
- E3 ubiquitin protein ligase TRIM69| HSD34| RFP like domain containing protein trimless| RFP-like domain-containing protein trimless| RING finger protein 36| RNF36| TRI69_HUMAN| Trif| Trim69| Trimless| Tripartite motif containing 69| tripartite motif containing protein 69| tripartite motif protein 69| Tripartite motif-containing protein 69
- E3 ubiquitin protein ligase TRIM69| HSD34| RFP like domain containing protein trimless| RFP-like domain-containing protein trimless| RING finger protein 36| RNF36| TRI69_HUMAN| Trif| Trim69| Trimless| Tripartite motif containing 69| tripartite motif containing protein 69| tripartite motif protein 69| Tripartite motif-containing protein 69
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86WT6
- Q86WT6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice