Anti-TSPAN4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: TSPAN4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-633
- 77803-633
- Antigen Symbol:
- TSPAN4
- TSPAN4
- Antigen Name:
- Tetraspanin-4 protein (107-201AA)
- Tetraspanin-4 protein (107-201AA)
- Antigen Synonyms:
- NAG 2|NAG-2|NAG2|Novel antigen 2|Tetraspan|Tetraspan TM4SF|Tetraspanin 4|Tetraspanin-4|Tetraspanin4|TM4SF7|Transmembrane 4 superfamily member 7|TSN4_HUMAN|TSPAN 4|Tspan-4|TSPAN4
- NAG 2|NAG-2|NAG2|Novel antigen 2|Tetraspan|Tetraspan TM4SF|Tetraspanin 4|Tetraspanin-4|Tetraspanin4|TM4SF7|Transmembrane 4 superfamily member 7|TSN4_HUMAN|TSPAN 4|Tspan-4|TSPAN4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14817
- O14817
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice