Anti-HS1BP3 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: HS1BP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-514
- 77827-513
- Antigen Symbol:
- HS1BP3
- HS1BP3
- Antigen Name:
- HCLS1-binding protein 3 protein (1-200AA)
- HCLS1-binding protein 3 protein (1-200AA)
- Antigen Synonyms:
- ETM2| FLJ14249| H1BP3_HUMAN| HCLS1 binding protein 3| HCLS1-binding protein 3| HS1 binding protein 3| HS1 BP3| HS1-binding protein 3| Hs1bp3| HSP1BP-3
- ETM2| FLJ14249| H1BP3_HUMAN| HCLS1 binding protein 3| HCLS1-binding protein 3| HS1 binding protein 3| HS1 BP3| HS1-binding protein 3| Hs1bp3| HSP1BP-3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53T59
- Q53T59
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice