Anti-USP17L5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP17L5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-253
- 77818-252
- Antigen Symbol:
- USP17L5
- USP17L5
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA)
- Ubiquitin carboxyl-terminal hydrolase 17-like protein 5 protein (19-235AA)
- Antigen Synonyms:
- USP17L5| Ubiquitin carboxyl-terminal hydrolase 17-like protein 5| EC 3.4.19.12| Deubiquitinating enzyme 17-like protein 5| Ubiquitin thioesterase 17-like protein 5| Ubiquitin-specific-processing protease 17-like protein 5
- USP17L5| Ubiquitin carboxyl-terminal hydrolase 17-like protein 5| EC 3.4.19.12| Deubiquitinating enzyme 17-like protein 5| Ubiquitin thioesterase 17-like protein 5| Ubiquitin-specific-processing protease 17-like protein 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A8MUK1
- A8MUK1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice