Anti-RCAN1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: RCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77819-985
- 77819-986
- Antigen Symbol:
- RCAN1
- RCAN1
- Antigen Name:
- Synthetic peptide of RCAN1
- Synthetic peptide of RCAN1
- Antigen Synonyms:
- Adapt 78|Adapt78|Calcipressin 1|Calcipressin-1|Calcium and oxidant inducible mRNA|CSP 1|CSP1|Down syndrome candidate region 1|Down syndrome critical region gene 1|Down syndrome critical region protein 1|DSC 1|DSC1|DSCR 1|DSCR1|MCIP 1|MCIP1|Modulatory calcineurin interacting protein 1|Myocyte enriched calcineurin interacting protein 1|Myocyte-enriched calcineurin-interacting protein 1|Near DSCR proline rich protein|RCAN1|RCAN1_HUMAN|RCN 1|RCN1|Regulator of calcineurin 1
- Adapt 78|Adapt78|Calcipressin 1|Calcipressin-1|Calcium and oxidant inducible mRNA|CSP 1|CSP1|Down syndrome candidate region 1|Down syndrome critical region gene 1|Down syndrome critical region protein 1|DSC 1|DSC1|DSCR 1|DSCR1|MCIP 1|MCIP1|Modulatory calcineurin interacting protein 1|Myocyte enriched calcineurin interacting protein 1|Myocyte-enriched calcineurin-interacting protein 1|Near DSCR proline rich protein|RCAN1|RCAN1_HUMAN|RCN 1|RCN1|Regulator of calcineurin 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53805
- P53805
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice