Anti-FOXQ1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FOXQ1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-260
- 77839-261
- Antigen Symbol:
- FOXQ1
- FOXQ1
- Antigen Name:
- Forkhead box protein Q1 protein (10-123AA)
- Forkhead box protein Q1 protein (10-123AA)
- Antigen Synonyms:
- Forkhead box protein Q1|Forkhead box Q1|FOX Q1|FOXQ 1|FOXQ1|FOXQ1_HUMAN|Hepatocyte nuclear factor 3 forkhead|Hepatocyte nuclear factor 3 forkhead homolog 1|HFH 1|HFH-1|HFH1|HNF 3/forkhead like protein 1|HNF-3/forkhead-like protein 1|HNF3/forkhead like protein 1|MGC77469|Q1|Winged helix / forkhead transcription factor|Winged helix/forkhead transcription factor|zgc:77469
- Forkhead box protein Q1|Forkhead box Q1|FOX Q1|FOXQ 1|FOXQ1|FOXQ1_HUMAN|Hepatocyte nuclear factor 3 forkhead|Hepatocyte nuclear factor 3 forkhead homolog 1|HFH 1|HFH-1|HFH1|HNF 3/forkhead like protein 1|HNF-3/forkhead-like protein 1|HNF3/forkhead like protein 1|MGC77469|Q1|Winged helix / forkhead transcription factor|Winged helix/forkhead transcription factor|zgc:77469
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9C009
- Q9C009
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice