About this item
Recommended dilution: WB: 1:2000 to 1:10000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RBM10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77796-769
- 77796-768
- Antigen Symbol:
- RBM10
- RBM10
- Antigen Name:
- RNA-binding protein 10 protein (501-700AA)
- RNA-binding protein 10 protein (501-700AA)
- Antigen Synonyms:
- DXS8237E|G patch domain containing protein 9|G patch domain-containing protein 9|GPATC9|GPATCH9|HGNC9896|KIAA0122|MGC1132|MGC997|RBM10_HUMAN|RNA binding motif protein 10|RNA binding protein 10|RNA binding protein S1-1|RNA-binding motif protein 10|RNA-binding protein 10|RNA-binding protein S1-1|S1-1|TARPS|ZRANB5
- DXS8237E|G patch domain containing protein 9|G patch domain-containing protein 9|GPATC9|GPATCH9|HGNC9896|KIAA0122|MGC1132|MGC997|RBM10_HUMAN|RNA binding motif protein 10|RNA binding protein 10|RNA binding protein S1-1|RNA-binding motif protein 10|RNA-binding protein 10|RNA-binding protein S1-1|S1-1|TARPS|ZRANB5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P98175
- P98175
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice