Anti-TYRO3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TYRO3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-787
- 77803-787
- Antigen Symbol:
- TYRO3
- TYRO3
- Antigen Name:
- Tyrosine-protein kinase receptor TYRO3 protein (728-873AA)
- Tyrosine-protein kinase receptor TYRO3 protein (728-873AA)
- Antigen Synonyms:
- Brt|BYK|DTK|Etk-2|protein tyrosine kinase 3|Rek|RSE|SKY|Tif|tyro3|TYRO3 protein tyrosine kinase|TYRO3_HUMAN|Tyrosine-protein kinase byk|Tyrosine-protein kinase DTK|Tyrosine-protein kinase receptor TYRO3|Tyrosine-protein kinase RSE|Tyrosine-protein kinase SKY|Tyrosine-protein kinase TIF
- Brt|BYK|DTK|Etk-2|protein tyrosine kinase 3|Rek|RSE|SKY|Tif|tyro3|TYRO3 protein tyrosine kinase|TYRO3_HUMAN|Tyrosine-protein kinase byk|Tyrosine-protein kinase DTK|Tyrosine-protein kinase receptor TYRO3|Tyrosine-protein kinase RSE|Tyrosine-protein kinase SKY|Tyrosine-protein kinase TIF
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q06418
- Q06418
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice