Anti-SARS Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: OXCT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77807-528
- 77807-529
- Antigen Symbol:
- SARS
- SARS
- Antigen Name:
- Serine--tRNA ligase, cytoplasmic protein (2-233AA)
- Serine--tRNA ligase, cytoplasmic protein (2-233AA)
- Antigen Synonyms:
- C78314|cytoplasmic|EC 6.1.1.11|FLJ36399|sarS|Sars1|Serine tRNA ligase 1, cytoplasmic|Serine tRNA ligase|Serine--tRNA ligase|serine--tRNA ligase, cytoplasmic|SerRS|SERS|Seryl tRNA Ser/Sec synthetase|Seryl tRNA synthetase|Seryl tRNA synthetase cytoplasmic|Seryl-tRNA Ser/Sec synthetase|Seryl-tRNA synthetase|seryl-tRNA synthetase, cytoplasmic|Seryl-tRNA(Ser/Sec) synthetase|Strs|SYSC_HUMAN
- C78314|cytoplasmic|EC 6.1.1.11|FLJ36399|sarS|Sars1|Serine tRNA ligase 1, cytoplasmic|Serine tRNA ligase|Serine--tRNA ligase|serine--tRNA ligase, cytoplasmic|SerRS|SERS|Seryl tRNA Ser/Sec synthetase|Seryl tRNA synthetase|Seryl tRNA synthetase cytoplasmic|Seryl-tRNA Ser/Sec synthetase|Seryl-tRNA synthetase|seryl-tRNA synthetase, cytoplasmic|Seryl-tRNA(Ser/Sec) synthetase|Strs|SYSC_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49591
- P49591
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together