Anti-ZSCAN32 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ZSCAN32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-466
- 77805-467
- Antigen Symbol:
- ZSCAN32
- ZSCAN32
- Antigen Name:
- Zinc finger and SCAN domain-containing protein 32 protein (6-248AA)
- Zinc finger and SCAN domain-containing protein 32 protein (6-248AA)
- Antigen Synonyms:
- ZSCAN32|ZNF434|HCCS5|Zinc finger and SCAN domain-containing protein 32|Human cervical cancer suppressor gene 5 protein|HCCS-5|Zinc finger protein 434
- ZSCAN32|ZNF434|HCCS5|Zinc finger and SCAN domain-containing protein 32|Human cervical cancer suppressor gene 5 protein|HCCS-5|Zinc finger protein 434
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NX65
- Q9NX65
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice