Anti-QPRT Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: QPRT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-474
- 77824-473
- Antigen Symbol:
- QPRT
- QPRT
- Antigen Name:
- Nicotinate-nucleotide pyrophosphorylase [carboxylating] protein (113-254AA)
- Nicotinate-nucleotide pyrophosphorylase [carboxylating] protein (113-254AA)
- Antigen Synonyms:
- NADC_HUMAN|Nicotinate nucleotide pyrophosphorylase (carboxylating)|Nicotinate-nucleotide pyrophosphorylase [carboxylating]|QAPRTase|QPRTase|Quinolinate phosphoribosyltransferase [decarboxylating]|Quinolinate phosphoribosyltransferase
- NADC_HUMAN|Nicotinate nucleotide pyrophosphorylase (carboxylating)|Nicotinate-nucleotide pyrophosphorylase [carboxylating]|QAPRTase|QPRTase|Quinolinate phosphoribosyltransferase [decarboxylating]|Quinolinate phosphoribosyltransferase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15274
- Q15274
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice