Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-DHX32 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-DHX32 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77830-220
  • 77830-219
  • Antigen Symbol:
  • DHX32
  • DHX32
  • Antigen Name:
  • Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA)
  • Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32 protein (301-500AA)
  • Antigen Synonyms:
  • 3110079L04Rik|AA408140|Ddx32|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32|DEAD/H box 32|DEAD/H helicase like protein 1|DEAD/H helicase-like protein 1|DEAH (Asp-Glu-Ala-His) box polypeptide 32|DEAH box protein 32|DHLP1|DHX32|DHX32_HUMAN|Helicase DDX32|HuDDX32|Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
  • 3110079L04Rik|AA408140|Ddx32|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 32|DEAD/H box 32|DEAD/H helicase like protein 1|DEAD/H helicase-like protein 1|DEAH (Asp-Glu-Ala-His) box polypeptide 32|DEAH box protein 32|DHLP1|DHX32|DHX32_HUMAN|Helicase DDX32|HuDDX32|Putative pre-mRNA-splicing factor ATP-dependent RNA helicase DHX32
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q7L7V1
  • Q7L7V1
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice