Anti-SLC1A2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC1A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-805
- 77799-806
- Antigen Symbol:
- SLC1A2
- SLC1A2
- Antigen Name:
- Excitatory amino acid transporter 2 protein (460-574AA)
- Excitatory amino acid transporter 2 protein (460-574AA)
- Antigen Synonyms:
- EAA2_HUMAN|EAAT2|Excitatory amino acid transporter 2|Excitotoxic amino acid transporter 2|Glial high affinity glutamate transporter|GLT 1|GLT1|Glutamate aspartate transporter II|Glutamate transporter 1|Glutamate/aspartate transporter II|Sodium dependent glutamate aspartate transporter 2|Sodium-dependent glutamate/aspartate transporter 2|solute carrier family 1 (glial high affinity glutamate transporter)|member 2|Solute carrier family 1 glial high affinity glutamate transporter member 2|Solute carrier family 1 member 2
- EAA2_HUMAN|EAAT2|Excitatory amino acid transporter 2|Excitotoxic amino acid transporter 2|Glial high affinity glutamate transporter|GLT 1|GLT1|Glutamate aspartate transporter II|Glutamate transporter 1|Glutamate/aspartate transporter II|Sodium dependent glutamate aspartate transporter 2|Sodium-dependent glutamate/aspartate transporter 2|solute carrier family 1 (glial high affinity glutamate transporter)|member 2|Solute carrier family 1 glial high affinity glutamate transporter member 2|Solute carrier family 1 member 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P43004
- P43004
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice