Anti-USP47 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: USP47
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-179
- 77834-178
- Antigen Symbol:
- USP47
- USP47
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 47 protein (1-157AA)
- Ubiquitin carboxyl-terminal hydrolase 47 protein (1-157AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN|USP47
- Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN|USP47
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96K76
- Q96K76
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice