Anti-USP6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77804-173
- 77804-172
- Antigen Symbol:
- USP6
- USP6
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 6 protein (1122-1359AA)
- Ubiquitin carboxyl-terminal hydrolase 6 protein (1122-1359AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 6|HRP1|Proto-oncogene TRE-2|TRE17|TRE2|Ubiquitin carboxyl-terminal hydrolase 6|Ubiquitin specific protease 6|Ubiquitin thiolesterase 6|Ubiquitin-specific-processing protease 6|UBP6_HUMAN|USP6
- Deubiquitinating enzyme 6|HRP1|Proto-oncogene TRE-2|TRE17|TRE2|Ubiquitin carboxyl-terminal hydrolase 6|Ubiquitin specific protease 6|Ubiquitin thiolesterase 6|Ubiquitin-specific-processing protease 6|UBP6_HUMAN|USP6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P35125
- P35125
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice