Anti-USP30 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP30
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-543
- 77831-544
- Antigen Symbol:
- USP30
- USP30
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 30 protein (101-400AA)
- Ubiquitin carboxyl-terminal hydrolase 30 protein (101-400AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 30|FLJ40511|Ub-specific protease 30|Ubiquitin carboxyl terminal hydrolase 30|Ubiquitin carboxyl-terminal hydrolase 30|Ubiquitin specific peptidase 30|Ubiquitin specific processing protease 30|Ubiquitin thioesterase 30|Ubiquitin thiolesterase 30|Ubiquitin-specific-processing protease 30|UBP30_HUMAN|usp30
- Deubiquitinating enzyme 30|FLJ40511|Ub-specific protease 30|Ubiquitin carboxyl terminal hydrolase 30|Ubiquitin carboxyl-terminal hydrolase 30|Ubiquitin specific peptidase 30|Ubiquitin specific processing protease 30|Ubiquitin thioesterase 30|Ubiquitin thiolesterase 30|Ubiquitin-specific-processing protease 30|UBP30_HUMAN|usp30
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q70CQ3
- Q70CQ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together