Anti-TAX1BP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TAX1BP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77807-727
- 77807-728
- Antigen Symbol:
- TAX1BP3
- TAX1BP3
- Antigen Name:
- Tax1-binding protein 3 protein (1-124AA)
- Tax1-binding protein 3 protein (1-124AA)
- Antigen Synonyms:
- Glutaminase interacting protein 3|Glutaminase-interacting protein 3|Tax interaction protein 1|Tax1 (human T cell leukemia virus type I) binding protein 3|Tax1 binding protein 3|Tax1-binding protein 3|TIP 1|TIP-1|TIP1|TX1B3_HUMAN
- Glutaminase interacting protein 3|Glutaminase-interacting protein 3|Tax interaction protein 1|Tax1 (human T cell leukemia virus type I) binding protein 3|Tax1 binding protein 3|Tax1-binding protein 3|TIP 1|TIP-1|TIP1|TX1B3_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14907
- O14907
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice