Anti-REXO4 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: REXO4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-028
- 77836-027
- Antigen Symbol:
- REXO4
- REXO4
- Antigen Name:
- RNA exonuclease 4 protein (1-422AA)
- RNA exonuclease 4 protein (1-422AA)
- Antigen Synonyms:
- Exonuclease XPMC2| hPMC2| Prevents mitotic catastrophe 2 protein homolog| REX4| REX4, RNA exonuclease 4 homolog| REX4, RNA exonuclease 4 homolog (S. cerevisiae)| rexo4| REXO4_HUMAN| RNA exonuclease 4| Xenopus prevents mitotic catatrophe 2 homolog| XPMC2| XPMC2 prevents mitotic catastrophe 2 homolog| XPMC2H
- Exonuclease XPMC2| hPMC2| Prevents mitotic catastrophe 2 protein homolog| REX4| REX4, RNA exonuclease 4 homolog| REX4, RNA exonuclease 4 homolog (S. cerevisiae)| rexo4| REXO4_HUMAN| RNA exonuclease 4| Xenopus prevents mitotic catatrophe 2 homolog| XPMC2| XPMC2 prevents mitotic catastrophe 2 homolog| XPMC2H
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZR2
- Q9GZR2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice