Anti-USP45 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP45
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-554
- 77829-555
- Antigen Symbol:
- USP45
- USP45
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 45 protein (1-240AA)
- Ubiquitin carboxyl-terminal hydrolase 45 protein (1-240AA)
- Antigen Synonyms:
- USP45|Ubiquitin carboxyl-terminal hydrolase 45|EC 3.4.19.12|Deubiquitinating enzyme 45|Ubiquitin thioesterase 45|Ubiquitin-specific-processing protease 45
- USP45|Ubiquitin carboxyl-terminal hydrolase 45|EC 3.4.19.12|Deubiquitinating enzyme 45|Ubiquitin thioesterase 45|Ubiquitin-specific-processing protease 45
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q70EL2
- Q70EL2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice