Anti-USP31 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: USP31
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-187
- 77830-188
- Antigen Symbol:
- USP31
- USP31
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 31 protein (1069-1196AA)
- Ubiquitin carboxyl-terminal hydrolase 31 protein (1069-1196AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 31|KIAA1203|Ubiquitin carboxyl terminal hydrolase 31|Ubiquitin carboxyl-terminal hydrolase 31|Ubiquitin specific peptidase 31|Ubiquitin specific processing protease 31|Ubiquitin specific protease 31|Ubiquitin specific proteinase 31|Ubiquitin thioesterase 31|Ubiquitin thiolesterase 31|Ubiquitin-specific-processing protease 31|UBP31_HUMAN|USP 31|USP31
- Deubiquitinating enzyme 31|KIAA1203|Ubiquitin carboxyl terminal hydrolase 31|Ubiquitin carboxyl-terminal hydrolase 31|Ubiquitin specific peptidase 31|Ubiquitin specific processing protease 31|Ubiquitin specific protease 31|Ubiquitin specific proteinase 31|Ubiquitin thioesterase 31|Ubiquitin thiolesterase 31|Ubiquitin-specific-processing protease 31|UBP31_HUMAN|USP 31|USP31
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q70CQ4
- Q70CQ4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice