Anti-CYP26C1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CYP26C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-407
- 77830-408
- Antigen Symbol:
- CYP26C1
- CYP26C1
- Antigen Name:
- Cytochrome P450 26C1 protein (331-460AA)
- Cytochrome P450 26C1 protein (331-460AA)
- Antigen Synonyms:
- CP26C_HUMAN|cyp26b1l|CYP26C1|cyp26d1|Cytochrome P450 26C1|Cytochrome P450 family 26 subfamily C polypeptide 1|Cytochrome P450|subfamily XXVIC polypeptide 1|EG546726|FFDD4|FLJ45301|MGC158120|retinoic acid catabolizing enzyme
- CP26C_HUMAN|cyp26b1l|CYP26C1|cyp26d1|Cytochrome P450 26C1|Cytochrome P450 family 26 subfamily C polypeptide 1|Cytochrome P450|subfamily XXVIC polypeptide 1|EG546726|FFDD4|FLJ45301|MGC158120|retinoic acid catabolizing enzyme
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6V0L0
- Q6V0L0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice