Anti-AP2S1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: AP2S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77828-237
- 77828-236
- Antigen Symbol:
- AP2S1
- AP2S1
- Antigen Name:
- Fusion protein of AP2S1
- Fusion protein of AP2S1
- Antigen Synonyms:
- Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP 2 subunit sigma|Adaptor protein complex AP-2 subunit sigma|Adaptor protein complex AP2 subunit sigma|Adaptor related protein complex 2 sigma 1 subunit|AI043088|AP 17|AP 2 complex subunit sigma 1|AP-2 complex subunit sigma|AP17|AP17 delta|AP2 complex subunit sigma 1|Ap2s1|AP2S1_HUMAN|CLAPS 2|CLAPS2|Clathrin adaptor complex AP2|sigma subunit|Clathrin adaptor protein AP17|Clathrin assembly protein 2 small chain|Clathrin associated/assembly/adaptor protein small 2|17-KD|Clathrin associated/assembly/adaptor protein|small 2 (17kD)|Clathrin coat assembly protein AP17|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|HA2 17 kDa subunit|MGC62945|Plasma membrane adaptor AP 2 17 kDa protein|Plasma membrane adaptor AP-2 17 kDa protein|Plasma membrane adaptor AP2 17 kDa protein|Sigma adaptin 3b|Sigma2 adaptin|Sigma2-adaptin
- Adapter-related protein complex 2 sigma subunit|Adaptor protein complex AP 2 subunit sigma|Adaptor protein complex AP-2 subunit sigma|Adaptor protein complex AP2 subunit sigma|Adaptor related protein complex 2 sigma 1 subunit|AI043088|AP 17|AP 2 complex subunit sigma 1|AP-2 complex subunit sigma|AP17|AP17 delta|AP2 complex subunit sigma 1|Ap2s1|AP2S1_HUMAN|CLAPS 2|CLAPS2|Clathrin adaptor complex AP2|sigma subunit|Clathrin adaptor protein AP17|Clathrin assembly protein 2 small chain|Clathrin associated/assembly/adaptor protein small 2|17-KD|Clathrin associated/assembly/adaptor protein|small 2 (17kD)|Clathrin coat assembly protein AP17|Clathrin coat associated protein AP17|Clathrin coat-associated protein AP17|HA2 17 kDa subunit|MGC62945|Plasma membrane adaptor AP 2 17 kDa protein|Plasma membrane adaptor AP-2 17 kDa protein|Plasma membrane adaptor AP2 17 kDa protein|Sigma adaptin 3b|Sigma2 adaptin|Sigma2-adaptin
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53680
- P53680
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together