Anti-REG1B Rabbit Polyclonal Antibody
Catalog # 77796-991
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Secretory pancreatic stone protein 2
- Antigen Symbol:REG1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P48304
- Antigen Synonyms:Lithostathine 1 beta|Lithostathine 1 beta precursor|Lithostathine-1-beta|Pancreatic stone protein 2|PSP 2|PSP-2|PSPS2|REG 1 beta|REG-1-beta|REG1B|REG1B_HUMAN|Regenerating islet derived 1 beta (pancreatic stone protein pancreatic thread protein)|Regenerating islet derived 1 beta|Regenerating islet derived protein 1 beta|Regenerating islet-derived protein 1-beta|Regenerating protein I beta|REGH|REGI BETA|REGL|Secretory pancreatic stone protein 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: REG1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human