Anti-USP6 Rabbit Recombinant Polyclonal Antibody
Catalog # 77804-169
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 6 protein (1122-1359AA)
- Antigen Symbol:USP6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P35125
- Antigen Synonyms:Deubiquitinating enzyme 6|HRP1|Proto-oncogene TRE-2|TRE17|TRE2|Ubiquitin carboxyl-terminal hydrolase 6|Ubiquitin specific protease 6|Ubiquitin thiolesterase 6|Ubiquitin-specific-processing protease 6|UBP6_HUMAN|USP6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: USP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat