Anti-TREML2 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TREML2
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-753
- 77828-754
- Antigen Symbol:
- TREML2
- TREML2
- Antigen Name:
- Trem-like transcript 2 protein (19-268AA)
- Trem-like transcript 2 protein (19-268AA)
- Antigen Synonyms:
- TREML2|C6orf76|TLT2|UNQ6268/PRO20473|Trem-like transcript 2 protein|TLT-2|Triggering receptor expressed on myeloid cells-like protein 2
- TREML2|C6orf76|TLT2|UNQ6268/PRO20473|Trem-like transcript 2 protein|TLT-2|Triggering receptor expressed on myeloid cells-like protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T2D2
- Q5T2D2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice