Anti-SAP30BP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SAP30BP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-428
- 77841-429
- Antigen Symbol:
- SAP30BP
- SAP30BP
- Antigen Name:
- SAP30-binding protein (1-308AA)
- SAP30-binding protein (1-308AA)
- Antigen Synonyms:
- HCNGP|HSV 1 binding|HSV-1 binding transcriptional regulator protein|HSV1 binding|HTRG|HTRP|Human transcriptional regulator protein|S30BP_HUMAN|SAP 30 binding protein|SAP30 binding protein|SAP30-binding protein|SAP30BP|Transcriptional regulator protein HCNGP
- HCNGP|HSV 1 binding|HSV-1 binding transcriptional regulator protein|HSV1 binding|HTRG|HTRP|Human transcriptional regulator protein|S30BP_HUMAN|SAP 30 binding protein|SAP30 binding protein|SAP30-binding protein|SAP30BP|Transcriptional regulator protein HCNGP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHR5
- Q9UHR5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice