Anti-SMYD3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: SMYD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-074
- 77842-073
- Antigen Symbol:
- SMYD3
- SMYD3
- Antigen Name:
- Histone-lysine N-methyltransferase SMYD3 protein (199-428AA)
- Histone-lysine N-methyltransferase SMYD3 protein (199-428AA)
- Antigen Synonyms:
- bA74P14.1 (novel protein)|bA74P14.1|FLJ21080|histone lysine N methyltransferase SMYD3|KMT3E|MGC104324|SET and MYND domain containing 3|SET and MYND domain containing protein 3|SET and MYND domain-containing protein 3|SMYD 3|Smyd3|SMYD3 protein|SMYD3_HUMAN|Zinc finger MYND domain containing 1|Zinc finger MYND domain containing protein 1|Zinc finger MYND domain-containing protein 1|Zinc finger protein subfamily 3A MYND domain containing 1|Zinc finger protein, subfamily 3A (MYND domain containing), 1|ZMYND 1|ZMYND1|ZNFN3A1
- bA74P14.1 (novel protein)|bA74P14.1|FLJ21080|histone lysine N methyltransferase SMYD3|KMT3E|MGC104324|SET and MYND domain containing 3|SET and MYND domain containing protein 3|SET and MYND domain-containing protein 3|SMYD 3|Smyd3|SMYD3 protein|SMYD3_HUMAN|Zinc finger MYND domain containing 1|Zinc finger MYND domain containing protein 1|Zinc finger MYND domain-containing protein 1|Zinc finger protein subfamily 3A MYND domain containing 1|Zinc finger protein, subfamily 3A (MYND domain containing), 1|ZMYND 1|ZMYND1|ZNFN3A1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7B4
- Q9H7B4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice