Anti-SLC39A6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SLC39A6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-316
- 77824-315
- Antigen Symbol:
- SLC39A6
- SLC39A6
- Antigen Name:
- Zinc transporter ZIP6 protein (170-320AA)
- Zinc transporter ZIP6 protein (170-320AA)
- Antigen Synonyms:
- Estrogen regulated protein LIV 1 6|Estrogen-regulated protein LIV-1|LIV 1|LIV 1 protein, estrogen regulated|S39A6_HUMAN|Solute carrier family 39 (metal ion transporter), member 6|solute carrier family 39 (zinc transporter), member 6|Solute carrier family 39 member 6|Zinc transporter ZIP6|ZIP 6|ZIP-6|Zrt and Irt like protein 6|Zrt- and Irt-like protein 6
- Estrogen regulated protein LIV 1 6|Estrogen-regulated protein LIV-1|LIV 1|LIV 1 protein, estrogen regulated|S39A6_HUMAN|Solute carrier family 39 (metal ion transporter), member 6|solute carrier family 39 (zinc transporter), member 6|Solute carrier family 39 member 6|Zinc transporter ZIP6|ZIP 6|ZIP-6|Zrt and Irt like protein 6|Zrt- and Irt-like protein 6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13433
- Q13433
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice