Anti-PCBD1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PCBD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77793-324
- 77793-325
- Antigen Symbol:
- PCBD1
- PCBD1
- Antigen Name:
- Pterin-4-alpha-carbinolamine dehydratase protein (2-104AA)
- Pterin-4-alpha-carbinolamine dehydratase protein (2-104AA)
- Antigen Synonyms:
- 4 alpha hydroxy tetrahydropterin dehydratase|4-alpha-hydroxy-tetrahydropterin dehydratase|6 pyruvoyl tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1)|6 pyruvoyl tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha|DCoH|Dimerization cofactor of hepatic nuclear factor 1 alpha|Dimerization cofactor of hepatocyte nuclear factor 1 alpha|Dimerization cofactor of hepatocyte nuclear factor 1-alpha|Dimerization cofactor of HNF1|Dimerizing cofactor for HNF1|PCBD 1|PCBD|PCBD1|PCD|Phenylalanine hydroxylase stimulating protein|Phenylalanine hydroxylase-stimulating protein|PHS|PHS_HUMAN|Pterin 4 alpha carbinolamine dehydratase|Pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1)|Pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha|Pterin carbinolamine dehydratase|Pterin-4-alpha-carbinolamine dehydratase
- 4 alpha hydroxy tetrahydropterin dehydratase|4-alpha-hydroxy-tetrahydropterin dehydratase|6 pyruvoyl tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1)|6 pyruvoyl tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha|DCoH|Dimerization cofactor of hepatic nuclear factor 1 alpha|Dimerization cofactor of hepatocyte nuclear factor 1 alpha|Dimerization cofactor of hepatocyte nuclear factor 1-alpha|Dimerization cofactor of HNF1|Dimerizing cofactor for HNF1|PCBD 1|PCBD|PCBD1|PCD|Phenylalanine hydroxylase stimulating protein|Phenylalanine hydroxylase-stimulating protein|PHS|PHS_HUMAN|Pterin 4 alpha carbinolamine dehydratase|Pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1)|Pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha|Pterin carbinolamine dehydratase|Pterin-4-alpha-carbinolamine dehydratase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61457
- P61457
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice