Anti-CBY1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: CBY1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77810-074
- 77810-075
- Antigen Symbol:
- CBY1
- CBY1
- Antigen Name:
- Fusion protein of CBY1
- Fusion protein of CBY1
- Antigen Synonyms:
- ARPP-binding protein|C22orf2|CBY|Cby1|CBY1_HUMAN|Cytosolic leucine rich protein|Cytosolic leucine-rich protein|Golgi and endoplasmic reticulum-associated 1|HRIHFB2025|PIGEA 14|PIGEA-14|PKD2 interactor|PKD2 interactor, Golgi and endoplasmic reticulum-associated 1|Protein Chibby|Protein chibby homolog 1
- ARPP-binding protein|C22orf2|CBY|Cby1|CBY1_HUMAN|Cytosolic leucine rich protein|Cytosolic leucine-rich protein|Golgi and endoplasmic reticulum-associated 1|HRIHFB2025|PIGEA 14|PIGEA-14|PKD2 interactor|PKD2 interactor, Golgi and endoplasmic reticulum-associated 1|Protein Chibby|Protein chibby homolog 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3M2
- Q9Y3M2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice