Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GPR82 Rabbit Recombinant Polyclonal Antibody
Anti-GPR82 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GPR82 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77834-262
  • 77834-261
  • Antigen Symbol:
  • GPR82
  • GPR82
  • Antigen Name:
  • Probable G-protein coupled receptor 82 protein (273-336AA)
  • Probable G-protein coupled receptor 82 protein (273-336AA)
  • Antigen Synonyms:
  • GPR82Probable G-protein coupled receptor 82
  • GPR82Probable G-protein coupled receptor 82
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96P67
  • Q96P67
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications