Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP9X Rabbit Recombinant Polyclonal Antibody
Anti-USP9X Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP9X Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77833-868
  • 77833-869
  • Antigen Symbol:
  • USP9X
  • USP9X
  • Antigen Name:
  • Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
  • Probable ubiquitin carboxyl-terminal hydrolase FAF-X protein (2360-2484AA)
  • Antigen Synonyms:
  • Deubiquitinating enzyme FAF X|Deubiquitinating enzyme FAF-X|DFFRX|Drosophila fat facets related X linked|FAF|Fafl|Fam|Fat facets homolog|Fat facets in mammals|Fat facets protein related X linked|Fat facets protein related, X-linked|Fat facets protein-related|hFAM|MRX99|Probable ubiquitin carboxyl terminal hydrolase FAF X|Probable ubiquitin carboxyl-terminal hydrolase FAF-X|Ubiquitin carboxyl-terminal hydrolase FAM|Ubiquitin specific peptidase 9 X linked|Ubiquitin specific peptidase 9, X-linked|Ubiquitin specific processing protease FAF X|Ubiquitin specific protease 9 X chromosome|Ubiquitin thioesterase FAF X|Ubiquitin thiolesterase FAF X|Ubiquitin thiolesterase FAF-X|Ubiquitin-specific protease 9|Ubiquitin-specific-processing protease FAF-X|USP9 (gene name)|Usp9x|_HUMAN|Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)|X chromosome|X-linked
  • Deubiquitinating enzyme FAF X|Deubiquitinating enzyme FAF-X|DFFRX|Drosophila fat facets related X linked|FAF|Fafl|Fam|Fat facets homolog|Fat facets in mammals|Fat facets protein related X linked|Fat facets protein related, X-linked|Fat facets protein-related|hFAM|MRX99|Probable ubiquitin carboxyl terminal hydrolase FAF X|Probable ubiquitin carboxyl-terminal hydrolase FAF-X|Ubiquitin carboxyl-terminal hydrolase FAM|Ubiquitin specific peptidase 9 X linked|Ubiquitin specific peptidase 9, X-linked|Ubiquitin specific processing protease FAF X|Ubiquitin specific protease 9 X chromosome|Ubiquitin thioesterase FAF X|Ubiquitin thiolesterase FAF X|Ubiquitin thiolesterase FAF-X|Ubiquitin-specific protease 9|Ubiquitin-specific-processing protease FAF-X|USP9 (gene name)|Usp9x|_HUMAN|Uubiquitin specific protease 9, X chromosome (fat facets like Drosophila)|X chromosome|X-linked
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q93008
  • Q93008
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications