Anti-DIXDC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: DIXDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77824-509
- 77824-508
- Antigen Symbol:
- CCD1;Ccd1Aalpha1L;Ccd1Abeta1L;Ccd1BalphaL;Ccd1BbetaL;Ccd1CL;Coiled coil DIX1;Coiled coil protein DIX1;Coiled-coil protein DIX1;Coiled-coil-DIX1;DIX domain containing 1;DIX domain containing protein 1;DIX domain-containing protein 1;DIXC1_HUMAN;DIXDC1;Dixin;KIAA1735
- CCD1;Ccd1Aalpha1L;Ccd1Abeta1L;Ccd1BalphaL;Ccd1BbetaL;Ccd1CL;Coiled coil DIX1;Coiled coil protein DIX1;Coiled-coil protein DIX1;Coiled-coil-DIX1;DIX domain containing 1;DIX domain containing protein 1;DIX domain-containing protein 1;DIXC1_HUMAN;DIXDC1;Dixin;KIAA1735
- Antigen Name:
- Dixin protein (70-333AA)
- Dixin protein (70-333AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q155Q3
- Q155Q3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice